LIF509444789 Life Chemicals Inc.

4,6-dibromo-2-methoxypyridin-3-amine

  • MDL number MFCD29999493
  • Cas Number : 1949816-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart