LIF509444794 Life Chemicals Inc.

acridin-9-ylmethanamine dihydrochloride

  • MDL number MFCD29999619
  • Cas Number : 93716-65-5
  • Molecular Weight : 281.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart