LIF509444802 Life Chemicals Inc.

3-(pyridin-3-yl)morpholine hydrochloride

  • MDL number MFCD30005379
  • Cas Number : 1949816-24-3
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart