LIF509444834 Life Chemicals Inc.

6-ethoxypyrimidin-4-amine

  • MDL number MFCD19205984
  • Cas Number : 3289-43-8
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart