LIF509444858 Life Chemicals Inc.

3-(piperazin-1-yl)-6-(1H-1,2,4-triazol-1-yl)pyridazine

  • MDL number MFCD28369863
  • Cas Number : 1788589-62-7
  • Molecular Weight : 231.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart