LIF509444930 Life Chemicals Inc.

3-(imidazo[1,2-a]pyrimidin-2-yl)aniline

  • MDL number MFCD03012714
  • Cas Number : 439108-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart