LIF509444933 Life Chemicals Inc.

6-(4-ethoxyphenyl)pyridazin-3(2H)-one

  • MDL number MFCD12143121
  • Cas Number : 95117-76-3
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart