LIF509444943 Life Chemicals Inc.

Ethyl 4-chlorophenylacethylacetate

  • MDL number MFCD03844412
  • Cas Number : 62088-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart