LIF509444953 Life Chemicals Inc.

2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonate potassium

  • MDL number MFCD00023093
  • Cas Number : 91893-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart