LIF509444954 Life Chemicals Inc.

4-isopropyl-2-piperazin-1-yl-1,3-benzothiazole dihydrochloride

  • MDL number MFCD27937786
  • Cas Number : 1864073-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart