LIF509445017 Life Chemicals Inc.

2-(3,3-difluorocyclobutyl)acetic acid

  • MDL number MFCD22199410
  • Cas Number : 1373503-48-0
  • Molecular Weight : 150.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart