LIF509445022 Life Chemicals Inc.

7-amino-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-5-ol hydrochloride

  • MDL number MFCD28359740
  • Cas Number : 1949816-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart