LIF509445095 Life Chemicals Inc.

2-(((4-fluorophenyl)thio)methyl)-5-hydroxy-4H-pyran-4-one

  • MDL number MFCD25963797
  • Cas Number : 500861-19-8
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart