LIF509445102 Life Chemicals Inc.

3,8-dibromo-1,6-naphthyridine

  • MDL number MFCD05155260
  • Cas Number : 17965-75-2
  • Molecular Weight : 287.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart