LIF509445122 Life Chemicals Inc.

4-[(1-methyl-1H-imidazol-2-yl)sulfonyl]piperidine hydrochloride

  • MDL number MFCD26792982
  • Cas Number : 1823885-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart