LIF509445125 Life Chemicals Inc.

1-(1,3-benzoxazol-2-yl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD12903230
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart