LIF509445168 Life Chemicals Inc.

4-(2H-1,2,3-triazol-2-yl)piperidine hydrochloride

  • MDL number MFCD11865131
  • Cas Number : 690261-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart