LIF509445181 Life Chemicals Inc.

6-(furan-2-yl)pyridine-3-carbonitrile

  • MDL number MFCD09025879
  • Cas Number : 619334-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart