LIF509445196 Life Chemicals Inc.

3-(pyrrolidin-3-ylmethyl)pyridine dihydrochloride

  • MDL number MFCD03840071
  • Cas Number : 1864064-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart