LIF509445203 Life Chemicals Inc.

1-(pyridin-2-yl)pyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD28388754
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart