LIF509445205 Life Chemicals Inc.

(2-aminoethoxy)cyclobutane hydrochloride

  • MDL number MFCD28148279
  • Cas Number : 1949816-30-1
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart