LIF509445207 Life Chemicals Inc.

2,3-dibromo-1,8-naphthyridine

  • MDL number MFCD06254515
  • Cas Number : 52626-31-0
  • Molecular Weight : 287.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart