LIF509445228 Life Chemicals Inc.

1-(prop-2-yn-1-yl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD11695061
  • Cas Number : 104501-02-2
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart