LIF509445230 Life Chemicals Inc.

2,2-dimethyl-3-(phenylamino)propan-1-ol

  • MDL number MFCD20662655
  • Cas Number : 94844-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart