LIF509445244 Life Chemicals Inc.

3,4-Dimethyl benzene sulfonyl chloride

  • MDL number MFCD00625742
  • Cas Number : 2905-30-8
  • Molecular Weight : 204.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart