LIF509445271 Life Chemicals Inc.

Methyl 6-Hydroxy-2-naphthoate

  • MDL number MFCD00100526
  • Cas Number : 17295-11-3
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart