LIF509445301 Life Chemicals Inc.

1-amino-2-biphenyl-4-ylpropan-2-ol

  • MDL number MFCD14529240
  • Cas Number : 1216165-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart