LIF509445315 Life Chemicals Inc.

2-amino-1-cyclopropyl-1-phenylethanol

  • MDL number MFCD17219202
  • Cas Number : 1268064-39-6
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart