LIF509445317 Life Chemicals Inc.

1-amino-2-(furan-2-yl)propan-2-ol

  • MDL number MFCD20441167
  • Cas Number : 933726-05-7
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart