LIF509445318 Life Chemicals Inc.

4-(aminomethyl)chroman-4-ol

  • MDL number MFCD11577339
  • Cas Number : 146471-52-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart