LIF509445323 Life Chemicals Inc.

3-amino-1-(2,5-dimethylfuran-3-yl)propan-1-ol

  • MDL number MFCD21130897
  • Cas Number : 1447966-95-1
  • Molecular Weight : 169.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart