LIF509445398 Life Chemicals Inc.

N-(Chloroacetyl)4-aminoacetanilide

  • MDL number MFCD00186762
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart