LIF509445449 Life Chemicals Inc.

N-Methylbutylamine

  • MDL number MFCD00009426
  • Cas Number : 110-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart