LIF509445766 Life Chemicals Inc.

5-(1,3-benzodioxol-5-yl)-2-piperazin-1-yl-6H-1,3,4-thiadiazine dihydrochloride

  • MDL number MFCD16653417
  • Cas Number : 1351612-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart