LIF509446034 Life Chemicals Inc.

3-(oxolan-2-yl)propanoic acid

  • MDL number MFCD03412485
  • Cas Number : 935-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart