LIF509446038 Life Chemicals Inc.

2-(3,5-dimethyl-1-(pyridin-2-yl)-1H-pyrazol-4-yl)ethan-1-amine

  • MDL number MFCD11576893
  • Cas Number : 1177355-64-4
  • Molecular Weight : 216.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart