LIF509446049 Life Chemicals Inc.

2-((3-(pyridin-3-yl)-1,2,4-oxadiazol-5-yl)methyl)aniline

  • MDL number MFCD17356082
  • Cas Number : 1273824-82-0
  • Molecular Weight : 252.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart