LIF509446071 Life Chemicals Inc.

N-methyl-2-(phenylsulfonyl)ethan-1-amine

  • MDL number MFCD02089404
  • Cas Number : 404033-91-6
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart