LIF509446482 Life Chemicals Inc.

5-(chloromethyl)-3-ethyl-1,2,4-oxadiazole

  • MDL number MFCD08059891
  • Cas Number : 50737-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart