LIF509446485 Life Chemicals Inc.

4-(4-methyl-4H-1,2,4-triazol-3-yl)piperidine

  • MDL number MFCD09055373
  • Cas Number : 297172-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart