LIF509446514 Life Chemicals Inc.

1-[3-(chloromethyl)piperidin-1-yl]ethan-1-one

  • MDL number MFCD11612379
  • Cas Number : 1082844-32-3
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart