LIF509446551 Life Chemicals Inc.

2-chloro-3-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD07352255
  • Cas Number : 871548-31-1
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart