LIF509446571 Life Chemicals Inc.

2-(1-methyl-1H-pyrazol-3-yl)acetic acid hydrochloride

  • MDL number MFCD22209866
  • Cas Number : 1357353-48-0
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart