LIF509446602 Life Chemicals Inc.

3-(propan-2-yl)-1,3,7-triazaspiro[4.4]nonane-2,4-dione hydrochloride

  • MDL number MFCD27995643
  • Cas Number : 1779128-57-2
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart