LIF509446604 Life Chemicals Inc.

4-chloro-6-(4-ethyl-2-methylpiperazin-1-yl)pyrimidine

  • MDL number MFCD27995689
  • Cas Number : 1707361-49-6
  • Molecular Weight : 240.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart