LIF509446728 Life Chemicals Inc.

4,5,6,7-tetrahydro-2-benzofuran-1-carboxylic acid

  • MDL number MFCD20656551
  • Cas Number : 1378818-17-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart