LIF509446744 Life Chemicals Inc.

5-(propan-2-yl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07376161
  • Cas Number : 89006-96-2
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart