LIF509446753 Life Chemicals Inc.

8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD08272087
  • Cas Number : 68774-78-7
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart