LIF509446761 Life Chemicals Inc.

1-(2-methoxyethyl)-2,6-dimethylpiperazine

  • MDL number MFCD09894141
  • Cas Number : 1017120-41-0
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart