LIF509446781 Life Chemicals Inc.

1,1,3,3-tetramethyl-2,3-dihydro-1H-indene

  • MDL number MFCD27995656
  • Cas Number : 4834-33-7
  • Molecular Weight : 174.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart